Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nbn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138873.2 | NP_620228.1 | 3 | 323 | ||
XM_006237916.1 | XP_006237978.1 | 3 | 366 | |||
GenBank mRNA | AF218575.1 | - | 3 | 327 | ||
BC085700.1 | - | 3 | 291 | 7130978 |