Search
Search

PTPN23| Gene Symbol: PTPN23 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001304482.1 | NP_001291411.1 | 11 | 937 | ||
| NM_015466.3 | NP_056281.1 | 12 | 1065 | |||
| XM_005265031.2 | XP_005265088.2 | 12 | 1016 | |||
| GenBank mRNA | AB025194.1 | - | 12 | 1013 | ||
| AB040904.2 | - | 12 | 1065 | |||
| AF290614.1 | - | 12 | 1011 | |||
| AK055157.1 | - | 12 | 1036 | |||
| AK289502.1 | - | 12 | 1014 | |||
| AK295877.1 | - | 12 | 985 | |||
| AK295888.1 | - | 11 | 928 | |||
| AK299906.1 | - | 11 | 908 | |||
| BC089042.1 | - | 12 | 983 | 6579163 | ||