Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Carhsp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_152790.2 | NP_690003.1 | 2, 3 | 199 | ||
XM_006245750.2 | XP_006245812.1 | 2, 3 | 982 | |||
XM_006245753.2 | XP_006245815.1 | 2, 3 | 353 | |||
XM_008767479.1 | XP_008765701.1 | 2, 3 | 676 | |||
GenBank mRNA | AF115346.1 | - | 1, 2 | 169 | ||
BC071173.1 | - | 2, 3 | 199 | 7099252 |