Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TSPO SIRNA Assays ›
Gene Symbol:
Tspo
Gene Name:
translocator protein
Gene Aliases:
Bzrp, MBR, PTBZR02, Ptbzr, RATPTBZR02
Chromosome Location:
Chr.7: 124460358 - 124470610 on Build Rnor_6.0
UniGene ID:
Rn.1820
Species:
Rat
Species Specific ID (Flybase ID):
-
siRNA ID
59135
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Tspo
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_012515.2
NP_036647.1
3
307
GenBank mRNA
FQ228784.1
-
3
309
J05122.1
-
2
250
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Mouse: NM_009775, L17306, AK151421, AA472650, AK172384, and others.
Gene Ontology Categories:
Function(s)
Process(es)
steroid biosynthetic process
ion transport
chloride transport
lipid transport
chemical synaptic transmission
aging
glial cell migration
response to manganese ion
response to vitamin B1
positive regulation of necrotic cell death
peripheral nervous system axon regeneration
adrenal gland development
response to progesterone
negative regulation of tumor necrosis factor production
response to testosterone
response to drug
positive regulation of apoptotic process
negative regulation of nitric oxide biosynthetic process
response to pain
behavioral response to pain
response to axon injury
regulation of steroid biosynthetic process
positive regulation of mitochondrial depolarization
positive regulation of calcium ion transport
contact inhibition
positive regulation of glial cell proliferation
negative regulation of glial cell proliferation
cellular response to lipopolysaccharide
cellular response to zinc ion
cellular hypotonic response
positive regulation of reactive oxygen species metabolic process
androgen binding
protein binding
benzodiazepine receptor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-fz59j:80/100.66.79.31:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0