Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ntf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013184.3 | NP_037316.2 | 3 | 868 | ||
GenBank mRNA | S69323.1 | - | 3 | 868 |