Hamburger Menu Button
Thermo Fisher Scientific Logo
登入
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Tubes
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Clinical Genomics
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO 和 CRO 服務
    • CDMO 服務
    • 臨床試驗 CRO 服務
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • Register for an Account
    • How to Order
    • Instrument Support
    • Technical Support Centers
    • Learning Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • 快速訂購
          • 登入
            登入
            Don't have an account ? Create Account
            • 帳戶
            • 訂單
            • 訂製產品與專案
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other TRAF2 SIRNA Assays ›
          Gene Symbol:
          TRAF2
          Gene Name:
          TNF receptor associated factor 2
          Gene Aliases:
          MGC:45012, TRAP, TRAP3
          Chromosome Location:
          Chr.9: 136881776 - 136926615 on Build GRCh38
          UniGene ID:
          Hs.522506
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 5928

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: TRAF2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_021138.3 NP_066961.2 3 290
          XM_011518973.2 XP_011517275.1 4 594
          XM_011518974.2 XP_011517276.1 3 1195
          XM_011518975.2 XP_011517277.1 4 522
          XM_011518976.2 XP_011517278.1 4 563
          XM_011518977.2 XP_011517279.1 3 283
          XM_011518978.2 XP_011517280.1 3 1704
          XM_017015094.1 XP_016870583.1 3 338
          XM_017015095.1 XP_016870584.1 3 1048
          GenBank mRNA AK054686.1 - 3 277
          AK222961.1 - 3 287
          AK289722.1 - 4 406
          AK294607.1 - 3 287
          AK298370.1 - 3 254
          BC032410.1 - 2 276 5216790
          BC033810.1 - 3 278 5182942
          BC043492.1 - 3 290 5767794
          BC064662.1 - 2 285 6528158
          BM785733.1 - 3 290
          BX538160.1 - 3 307
          HM991672.1 - 2 233

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          scaffold/adaptor protein

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of T cell cytokine production
          protein complex assembly
          activation of cysteine-type endopeptidase activity involved in apoptotic process
          signal transduction
          I-kappaB kinase/NF-kappaB signaling
          activation of NF-kappaB-inducing kinase activity
          regulation of tumor necrosis factor-mediated signaling pathway
          regulation of necrotic cell death
          protein catabolic process
          positive regulation of interleukin-2 production
          tumor necrosis factor-mediated signaling pathway
          negative regulation of glial cell apoptotic process
          response to endoplasmic reticulum stress
          regulation of apoptotic process
          positive regulation of JUN kinase activity
          cellular protein complex assembly
          positive regulation of T cell activation
          regulation of immunoglobulin secretion
          positive regulation of sequence-specific DNA binding transcription factor activity
          positive regulation of NF-kappaB transcription factor activity
          protein heterooligomerization
          protein autoubiquitination
          intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
          protein homotrimerization
          protein K63-linked ubiquitination
          death-inducing signaling complex assembly
          cellular response to nitric oxide
          positive regulation of protein homodimerization activity
          activation of cysteine-type endopeptidase activity involved in apoptotic signaling pathway
          programmed necrotic cell death
          negative regulation of neuron death
          regulation of extrinsic apoptotic signaling pathway via death domain receptors
          negative regulation of extrinsic apoptotic signaling pathway via death domain receptors
          positive regulation of tumor necrosis factor-mediated signaling pathway
          positive regulation of I-kappaB phosphorylation
          positive regulation of extrinsic apoptotic signaling pathway
          ubiquitin-protein transferase activity
          signal transducer activity
          tumor necrosis factor receptor binding
          CD40 receptor binding
          protein binding
          zinc ion binding
          ligase activity
          enzyme binding
          protein kinase binding
          protein phosphatase binding
          mitogen-activated protein kinase kinase kinase binding
          ubiquitin protein ligase binding
          thioesterase binding
          protein complex binding
          identical protein binding
          sphingolipid binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Hong Kong flag icon
          Hong Kong

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0