Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cldn3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031700.2 | NP_113888.2 | 1 | 628 | ||
GenBank mRNA | BC062411.1 | - | 1 | 253 | 5623891 | |
M74067.1 | - | 1 | 348 |
Gene Symbol: Cldn4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001012022.1 | NP_001012022.1 | 1 | 382 | ||
GenBank mRNA | BC083582.1 | - | 1 | 382 | 7130270 |