Search
Search

Rara| Gene Symbol: Rara | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031528.2 | NP_113716.2 | 2 | 430 | ||
| XM_008767945.1 | XP_008766167.1 | 2 | 230 | |||
| GenBank mRNA | AJ002940.1 | - | 2 | 154 | ||
| AJ002941.1 | - | 2 | 170 | |||
| BC099830.1 | - | 2 | 430 | 7933868 | ||
| U15211.1 | - | 2 | 436 | |||