Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cst6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133566.1 | NP_598250.1 | 1 | 455 | ||
GenBank mRNA | AY046415.1 | - | 1 | 455 |