Search
Search

LOC100363178
Adora3| Gene Symbol: LOC100363178 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003749374.3 | XP_003749422.1 | 1064 | |||
| Gene Symbol: Adora3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF102804.1 | - | 1 | 242 | ||
| DQ075463.1 | - | 1 | 245 | |||
| M94152.1 | - | 560 | ||||
| X93219.1 | - | 1 | 245 | |||
Set Membership: |
|