siRNA Details
Gene Symbol: Hbg1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_172093.1 | NP_742090.1 | 2 | 130 | ||
GenBank mRNA | BC157810.1 | - | 2 | 164 | 9027502 | |
FQ221361.1 | - | 2 | 182 | |||
FQ222999.1 | - | 2 | 177 | |||
FQ228385.1 | - | 2 | 184 | |||
FQ228657.1 | - | 2 | 180 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
