siRNA Details
Gene Symbol: Chrna7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007390.3 | NP_031416.3 | 2 | 151 | ||
GenBank mRNA | AF225980.1 | - | 2 | 141 | ||
AK034208.1 | - | 2 | 195 | |||
AK157809.1 | - | 2 | 148 | |||
AY574266.1 | - | 2 | 101 | |||
L37663.1 | - | 2 | 151 |

More Information
Other Species Targeted

Panther Classification:
Molecular Function -






