Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other APC SIRNA Assays ›
          Gene Symbol:
          Apc
          Gene Name:
          adenomatosis polyposis coli
          Gene Aliases:
          AI047805, AU020952, AW124434, CC1, Min, mAPC
          Chromosome Location:
          Chr.18: 34220307 - 34322190 on Build GRCm38
          UniGene ID:
          Mm.384171
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 60085

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Apc
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_007462.3 NP_031488.2 3 475
          GenBank mRNA AK042021.1 - 3 476
          AK045053.1 - 3 446
          AK046280.1 - 3 466
          AK080907.1 - 3 475
          AK082724.1 - 3 221
          BC141141.1 - 2 188 9055753
          M88127.1 - 2 164

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_012499, XM_006254509, XM_006254511, XM_006254513, XM_006254520, and others.

          Gene Ontology Categories:

          Function(s) Process(es)

          mitotic cytokinesis
          kidney development
          hair follicle development
          protein complex assembly
          cellular response to DNA damage stimulus
          negative regulation of microtubule depolymerization
          cell cycle arrest
          metaphase/anaphase transition of mitotic cell cycle
          mitotic spindle assembly checkpoint
          establishment or maintenance of cell polarity
          pattern specification process
          axonogenesis
          negative regulation of cell proliferation
          axis specification
          anterior/posterior pattern specification
          dorsal/ventral pattern formation
          proximal/distal pattern formation
          Wnt signaling pathway
          cell migration
          stem cell population maintenance
          negative regulation of Wnt signaling pathway
          regulation of cell migration
          positive regulation of cell migration
          regulation of epithelial cell differentiation
          positive regulation of epithelial cell differentiation
          positive regulation of microtubule polymerization
          cytoplasmic microtubule organization
          neuron projection development
          positive regulation of pseudopodium assembly
          regulation of microtubule-based process
          T cell differentiation in thymus
          somatic stem cell population maintenance
          negative regulation of odontogenesis
          positive regulation of apoptotic process
          negative regulation of apoptotic process
          negative regulation of MAPK cascade
          skin development
          canonical Wnt signaling pathway involved in negative regulation of apoptotic process
          canonical Wnt signaling pathway involved in positive regulation of apoptotic process
          regulation of cell differentiation
          positive regulation of cell differentiation
          regulation of osteoblast differentiation
          regulation of osteoclast differentiation
          positive regulation of protein catabolic process
          negative regulation of cyclin-dependent protein serine/threonine kinase activity
          positive regulation of cell adhesion
          muscle cell cellular homeostasis
          thymus development
          negative regulation of epithelial cell proliferation
          regulation of nitrogen compound metabolic process
          chromosome organization
          regulation of cell cycle
          positive regulation of cell division
          regulation of attachment of spindle microtubules to kinetochore
          retina development in camera-type eye
          canonical Wnt signaling pathway
          negative regulation of epithelial cell proliferation involved in prostate gland development
          negative regulation of canonical Wnt signaling pathway
          regulation of glutamate metabolic process
          protease binding
          protein binding
          beta-catenin binding
          microtubule binding
          protein kinase regulator activity
          protein kinase binding
          protein complex binding
          gamma-catenin binding
          microtubule plus-end binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0