Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other APC SIRNA Assays ›
Gene Symbol:
Apc
Gene Name:
adenomatosis polyposis coli
Gene Aliases:
AI047805, AU020952, AW124434, CC1, Min, mAPC
Chromosome Location:
Chr.18: 34220307 - 34322190 on Build GRCm38
UniGene ID:
Mm.384171
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
60172
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Apc
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_007462.3
NP_031488.2
16
3706
GenBank mRNA
AK046280.1
-
16
3696
BC141141.1
-
15
3419
9055753
M88127.1
-
15
3395
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
mitotic cytokinesis
kidney development
hair follicle development
protein complex assembly
cellular response to DNA damage stimulus
negative regulation of microtubule depolymerization
cell cycle arrest
metaphase/anaphase transition of mitotic cell cycle
mitotic spindle assembly checkpoint
establishment or maintenance of cell polarity
pattern specification process
axonogenesis
negative regulation of cell proliferation
axis specification
anterior/posterior pattern specification
dorsal/ventral pattern formation
proximal/distal pattern formation
Wnt signaling pathway
cell migration
stem cell population maintenance
negative regulation of Wnt signaling pathway
regulation of cell migration
positive regulation of cell migration
regulation of epithelial cell differentiation
positive regulation of epithelial cell differentiation
positive regulation of microtubule polymerization
cytoplasmic microtubule organization
neuron projection development
positive regulation of pseudopodium assembly
regulation of microtubule-based process
T cell differentiation in thymus
somatic stem cell population maintenance
negative regulation of odontogenesis
positive regulation of apoptotic process
negative regulation of apoptotic process
negative regulation of MAPK cascade
skin development
canonical Wnt signaling pathway involved in negative regulation of apoptotic process
canonical Wnt signaling pathway involved in positive regulation of apoptotic process
regulation of cell differentiation
positive regulation of cell differentiation
regulation of osteoblast differentiation
regulation of osteoclast differentiation
positive regulation of protein catabolic process
negative regulation of cyclin-dependent protein serine/threonine kinase activity
positive regulation of cell adhesion
muscle cell cellular homeostasis
thymus development
negative regulation of epithelial cell proliferation
regulation of nitrogen compound metabolic process
chromosome organization
regulation of cell cycle
positive regulation of cell division
regulation of attachment of spindle microtubules to kinetochore
retina development in camera-type eye
canonical Wnt signaling pathway
negative regulation of epithelial cell proliferation involved in prostate gland development
negative regulation of canonical Wnt signaling pathway
regulation of glutamate metabolic process
protease binding
protein binding
beta-catenin binding
microtubule binding
protein kinase regulator activity
protein kinase binding
protein complex binding
gamma-catenin binding
microtubule plus-end binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-2rf75:80/100.66.79.83:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0