Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Speg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001085371.1 | NP_001078840.1 | 11 | 3038 | ||
NM_001173477.1 | NP_001166948.1 | 5 | 1051 | |||
GenBank mRNA | AK164360.1 | - | 11 | 3017 | ||
BC048698.1 | - | 5 | 948 | |||
BC062643.1 | - | 6 | 1163 | |||
U57098.1 | - | 4 | 878 |
Set Membership: |
G-protein coupled receptor |