siRNA Details
Gene Symbol: Baat | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007519.3 | NP_031545.2 | 4 | 1495 | ||
GenBank mRNA | AA255212.1 | - | 1 | 367 | ||
AK017923.1 | - | 4 | 1445 | |||
BC012683.1 | - | 4 | 1442 | |||
U95215.1 | - | 4 | 1403 |

More Information
Other Species Targeted

