Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cav3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007617.3 | NP_031643.1 | 1, 2 | 417 | ||
GenBank mRNA | BC024383.1 | - | 1, 2 | 427 | 4988598 | |
CJ057698.1 | - | 1, 2 | 194 | |||
U36579.1 | - | 1, 2 | 231 |