Search
Search

Cd9| Gene Symbol: Cd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007657.3 | NP_031683.1 | 4, 5 | 460 | ||
| GenBank mRNA | AK002251.1 | - | 4, 5 | 466 | ||
| AK012793.1 | - | 4, 5 | 466 | |||
| AK151443.1 | - | 4, 5 | 426 | |||
| AK151619.1 | - | 4, 5 | 413 | |||
| AK170310.1 | - | 4, 5 | 413 | |||
| BC070474.1 | - | 4, 5 | 460 | 30634238 | ||
| L08115.1 | - | 4, 5 | 398 | |||