Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GUCY2C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004963.3 | NP_004954.2 | 5, 6 | 867 | ||
XM_011520631.2 | XP_011518933.1 | 5, 6 | 765 | |||
GenBank mRNA | AK310034.1 | - | 5, 6 | 857 | ||
BC136544.1 | - | 5, 6 | 776 | 9020534 | ||
BC136545.1 | - | 5, 6 | 776 | 9020535 | ||
M73489.1 | - | 5, 6 | 778 | |||
S57551.1 | - | 5, 6 | 847 |