Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Col1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007743.2 | NP_031769.2 | 6 | 422 | ||
GenBank mRNA | AK031577.1 | - | 6 | 418 | ||
AK132496.1 | - | 6 | 418 | |||
AK157724.1 | - | 5 | 1067 | |||
AK159212.1 | - | 6 | 423 | |||
AK159408.1 | - | 6 | 419 | |||
AK159578.1 | - | 6 | 419 | |||
AK160098.1 | - | 6 | 422 | |||
AK160787.1 | - | 6 | 418 | |||
AK160788.1 | - | 6 | 417 | |||
AK160790.1 | - | 6 | 418 | |||
AK160946.1 | - | 6 | 421 | |||
AK161017.1 | - | 6 | 423 | |||
AK161605.1 | - | 6 | 418 | |||
AK164808.1 | - | 6 | 418 | |||
BC007158.1 | - | 6 | 402 | 3487477 | ||
BC042503.1 | - | 6 | 410 | 3586272 | ||
X58251.1 | - | 6 | 358 |