Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Reep5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007874.3 | NP_031900.3 | 4 | 515 | ||
GenBank mRNA | AK010507.1 | - | 4 | 467 | ||
AK013146.1 | - | 4 | 446 | |||
AK151058.1 | - | 4 | 470 | |||
AK151271.1 | - | 4 | 470 | |||
AK152361.1 | - | 4 | 444 | |||
AK152503.1 | - | 4 | 470 | |||
AK167385.1 | - | 4 | 444 | |||
AY562233.1 | - | 4 | 421 | |||
BC013052.1 | - | 4 | 446 | 3585290 | ||
BU053115.1 | - | 4 | 515 | |||
U28168.1 | - | 4 | 444 |