Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Who We Serve
Biotech
Biopharma
CDMO
Lab Diagnostics
Industrial and Applied Sciences
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other DMD SIRNA Assays ›
Gene Symbol:
Dmd
Gene Name:
dystrophin, muscular dystrophy
Gene Aliases:
DXSmh7, DXSmh9, Dp427, Dp71, dys, mdx, pke
Chromosome Location:
Chr.X: 82310713 - 85205050 on Build GRCm38
UniGene ID:
Mm.275608
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
61163
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
286.00
Your Price
Online offer:
277.65
277.65
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Dmd
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_007868.5
NP_031894.1
3, 4
430
GenBank mRNA
AK044536.1
-
3, 4
230
AK052856.1
-
3, 4
431
M68859.1
-
3, 4
403
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Human: NM_004006, NM_004009, NM_004010, NM_004007, NM_000109, and others.
This siRNA also targets the following transcripts in Rat: XM_008773281.
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
Gene Ontology Categories:
Function(s)
Process(es)
positive regulation of cell-matrix adhesion
regulation of heart rate
regulation of transcription, DNA-templated
muscle organ development
skeletal muscle tissue development
establishment of blood-nerve barrier
regulation of gene expression
regulation of release of sequestered calcium ion into cytosol by sarcoplasmic reticulum
regulation of cardiac muscle contraction by regulation of the release of sequestered calcium ion
positive regulation of neuron projection development
regulation of skeletal muscle contraction by regulation of release of sequestered calcium ion
regulation of skeletal muscle contraction
myotube cell development
muscle attachment
olfactory nerve structural organization
negative regulation of peptidyl-serine phosphorylation
cellular protein localization
response to muscle stretch
regulation of membrane potential
peptide biosynthetic process
cellular protein complex assembly
neurotransmitter receptor metabolic process
positive regulation of neuron differentiation
muscle cell cellular homeostasis
muscle fiber development
nucleus localization
cardiac muscle contraction
regulation of ryanodine-sensitive calcium-release channel activity
establishment of glial blood-brain barrier
negative regulation of ERK1 and ERK2 cascade
cardiac muscle cell action potential
regulation of cellular response to growth factor stimulus
regulation of voltage-gated calcium channel activity
negative regulation of peptidyl-cysteine S-nitrosylation
positive regulation of sodium ion transmembrane transporter activity
dystroglycan binding
actin binding
structural constituent of cytoskeleton
protein binding
zinc ion binding
structural constituent of muscle
myosin binding
vinculin binding
metal ion binding
nitric-oxide synthase binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0