Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PTK2 SIRNA Assays ›
          Gene Symbol:
          Ptk2
          Gene Name:
          PTK2 protein tyrosine kinase 2
          Gene Aliases:
          FAK, FRNK, Fadk, mKIAA4203, p125FAK
          Chromosome Location:
          Chr.15: 73205102 - 73423275 on Build GRCm38
          UniGene ID:
          Mm.254494
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 61445

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Ptk2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001130409.1 NP_001123881.1 5 798
          NM_007982.2 NP_032008.2 4 643
          GenBank mRNA AB011499.1 - 2 331
          AB030035.1 - 2 331
          AK005468.1 - 6 893
          AK079821.1 - 5 895
          AK220543.1 - 4 643
          BC030180.1 - 5 728
          JN971016.1 - 2 331
          M95408.1 - 3 440

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          MAPK cascade
          microtubule cytoskeleton organization
          angiogenesis
          blood vessel development
          vasculogenesis
          neuron migration
          regulation of protein phosphorylation
          positive regulation of protein phosphorylation
          protein phosphorylation
          movement of cell or subcellular component
          cell adhesion
          signal complex assembly
          epidermal growth factor receptor signaling pathway
          transforming growth factor beta receptor signaling pathway
          integrin-mediated signaling pathway
          JNK cascade
          multicellular organism development
          positive regulation of cell proliferation
          regulation of cell shape
          regulation of epithelial cell migration
          positive regulation of phosphatidylinositol 3-kinase signaling
          positive regulation of smooth muscle cell migration
          phosphorylation
          peptidyl-tyrosine phosphorylation
          central nervous system neuron axonogenesis
          negative regulation of cell-cell adhesion
          regulation of cell adhesion
          extracellular matrix organization
          positive regulation of cell growth
          positive regulation of cell migration
          negative regulation of cell migration
          cellular chloride ion homeostasis
          regulation of cell adhesion mediated by integrin
          peptidyl-tyrosine autophosphorylation
          establishment of nucleus localization
          regulation of cell proliferation
          negative regulation of apoptotic process
          endothelial cell migration
          innate immune response
          regulation of osteoblast differentiation
          positive regulation of cell adhesion
          positive regulation of protein kinase activity
          positive regulation of vasodilation
          negative regulation of organ growth
          protein autophosphorylation
          ephrin receptor signaling pathway
          positive regulation of smooth muscle cell proliferation
          positive regulation of phagocytosis
          negative regulation of axonogenesis
          positive regulation of synaptic transmission
          regulation of focal adhesion assembly
          positive regulation of protein kinase B signaling
          negative regulation of synapse assembly
          positive regulation of glial cell proliferation
          growth hormone receptor signaling pathway
          cellular response to transforming growth factor beta stimulus
          regulation of substrate adhesion-dependent cell spreading
          positive regulation of protein ubiquitination involved in ubiquitin-dependent protein catabolic process
          negative regulation of anoikis
          nucleotide binding
          actin binding
          protein kinase activity
          protein tyrosine kinase activity
          non-membrane spanning protein tyrosine kinase activity
          signal transducer activity
          receptor binding
          integrin binding
          protein binding
          ATP binding
          JUN kinase binding
          kinase activity
          transferase activity
          transferase activity, transferring phosphorus-containing groups
          protein kinase binding
          protein complex binding
          SH2 domain binding
          identical protein binding
          phosphatidylinositol 3-kinase binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2025 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-green-55658555d6-8mgq9:80/100.66.76.204:80.
          git-commit: ec8e7df5f8fec8d765bd419205f1b5046a016d37
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.41.0-Offline