Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcm2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008104.2 | NP_032130.2 | 1, 2 | 132 | ||
GenBank mRNA | AF081556.1 | - | 1, 2 | 98 | ||
AK135689.1 | - | 2, 3 | 238 | |||
BC110631.1 | - | 1, 2 | 162 | 40054293 | ||
BC110632.1 | - | 1, 2 | 162 | 40054295 | ||
D88611.1 | - | 1, 2 | 339 |