Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other FOXM1 SIRNA Assays ›
          Gene Symbol:
          FOXM1
          Gene Name:
          forkhead box M1
          Gene Aliases:
          FKHL16, FOXM1B, HFH-11, HFH11, HNF-3, INS-1, MPHOSPH2, MPP-2, MPP2, PIG29, TRIDENT
          Chromosome Location:
          Chr.12: 2857681 - 2877155 on Build GRCh38
          UniGene ID:
          Hs.239
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 6220

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: FOXM1
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001243088.1 NP_001230017.1 2 408
          NM_001243089.1 NP_001230018.1 2 408
          NM_021953.3 NP_068772.2 2 408
          NM_202002.2 NP_973731.1 2 408
          NM_202003.2 NP_973732.1 2 408
          XM_005253676.3 XP_005253733.1 2 408
          XM_011520930.2 XP_011519232.1 2 408
          XM_011520931.2 XP_011519233.1 2 408
          XM_011520932.1 XP_011519234.1 2 408
          XM_011520933.1 XP_011519235.1 2 408
          XM_011520934.2 XP_011519236.1 2 408
          XM_011520935.1 XP_011519237.1 2 408
          GenBank mRNA AK291206.1 - 2 311
          AK313845.1 - 2 254
          AY542306.1 - 1 176
          BC006192.2 - 2 293 3833837
          BC006529.2 - 2 293 3834244
          BC012863.1 - 1 144 3881055
          BQ229497.1 - 2 392
          BT006986.1 - 1 125
          DB137846.1 - 2 410
          U74612.1 - 2 239
          U74613.1 - 2 232
          U83113.1 - 2 194

          Back To Top

          More Information


          Gene Ontology Categories:

          Function(s) Process(es)

          G2/M transition of mitotic cell cycle
          negative regulation of transcription from RNA polymerase II promoter
          DNA repair
          transcription, DNA-templated
          DNA damage response, signal transduction by p53 class mediator resulting in transcription of p21 class mediator
          positive regulation of cell proliferation
          negative regulation of stress-activated MAPK cascade
          response to gonadotropin
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          regulation of Ras protein signal transduction
          response to cAMP
          regulation of cell cycle
          regulation of cell cycle arrest
          negative regulation of cell aging
          regulation of reactive oxygen species metabolic process
          positive regulation of double-strand break repair
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          protein binding
          protein kinase binding
          sequence-specific DNA binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Responsibility Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          México flag icon
          México

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0