siRNA Details
Gene Symbol: Hmgb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008252.3 | NP_032278.1 | 3 | 413 | ||
GenBank mRNA | AK003773.1 | - | 3 | 319 | ||
AK008443.1 | - | 3 | 324 | |||
AK011467.1 | - | 3 | 320 | |||
AK012568.1 | - | 3 | 380 | |||
AK076075.1 | - | 3 | 349 | |||
AK135296.1 | - | 3 | 319 | |||
AK135297.1 | - | 3 | 319 | |||
AK146212.1 | - | 3 | 348 | |||
AK151164.1 | - | 3 | 327 | |||
AK153858.1 | - | 3 | 322 | |||
BC002050.1 | - | 3 | 293 | 3489780 | ||
BC046759.1 | - | 3 | 317 | 5693598 | ||
BC083108.1 | - | 3 | 311 | 5320702 | ||
X67668.1 | - | 3 | 361 | |||
Z46757.1 | - | 3 | 322 |

More Information
Other Species Targeted

Panther Classification:
