siRNA Details
Gene Symbol: Hoxb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008269.1 | NP_032295.1 | 2 | 623 | ||
GenBank mRNA | BC016893.1 | - | 2 | 623 | 4223365 | |
X56461.1 | - | 2 | 1136 |

More Information
Other Species Targeted

Panther Classification:
