Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hnf4a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008261.2 | NP_032287.2 | 3, 4 | 506 | ||
GenBank mRNA | AK143948.1 | - | 3, 4 | 531 | ||
BC039220.1 | - | 3, 4 | 506 | 4238842 | ||
D29015.1 | - | 3, 4 | 433 | |||
EF193391.1 | - | 3, 4 | 305 | |||
EF193393.1 | - | 3, 4 | 331 | |||
FJ608819.1 | - | 4, 5 | 460 | |||
FJ608820.1 | - | 4, 5 | 460 | |||
FJ608821.1 | - | 4, 5 | 460 |