Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kcnc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008421.3 | NP_032447.1 | 2 | 2722 | ||
GenBank mRNA | AK086710.1 | - | 1 | 404 | ||
AK147476.1 | - | 2 | 2695 | |||
AK147682.1 | - | 2 | 2695 | |||
BC125470.1 | - | 2 | 1559 | 40129985 | ||
BC132439.1 | - | 2 | 1559 | 40130716 | ||
Y07521.1 | - | 2 | 1648 |