Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Promotions
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Order Status
Quick Order
Promos
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other TCF7L2 SIRNA Assays ›
Gene Symbol:
TCF7L2
Gene Name:
transcription factor 7 like 2
Gene Aliases:
TCF-4, TCF4
Chromosome Location:
Chr.10: 112950219 - 113167678 on Build GRCh38
UniGene ID:
Hs.593995
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
6268
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: TCF7L2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001146274.1
NP_001139746.1
1
631
NM_001146283.1
NP_001139755.1
1
631
NM_001146284.1
NP_001139756.1
1
631
NM_001146285.1
NP_001139757.1
1
631
NM_001146286.1
NP_001139758.1
1
631
NM_001198525.1
NP_001185454.1
1
631
NM_001198526.1
NP_001185455.1
1
631
NM_001198527.1
NP_001185456.1
1
631
NM_001198528.1
NP_001185457.1
1
631
NM_001198529.1
NP_001185458.1
1
631
NM_001198530.1
NP_001185459.1
1
631
NM_001198531.1
NP_001185460.1
1
631
NM_030756.4
NP_110383.2
1
631
XM_005270071.1
XP_005270128.1
1
662
XM_005270075.1
XP_005270132.1
1
662
XM_005270077.1
XP_005270134.1
1
662
XM_005270078.1
XP_005270135.1
1
662
XM_005270079.1
XP_005270136.1
1
662
XM_005270080.1
XP_005270137.1
1
662
XM_005270084.1
XP_005270141.1
1
662
XM_005270085.1
XP_005270142.1
1
662
XM_005270086.1
XP_005270143.1
1
662
XM_005270088.1
XP_005270145.1
1
662
XM_005270089.1
XP_005270146.1
1
662
XM_005270091.2
XP_005270148.1
1
662
XM_005270092.1
XP_005270149.1
1
662
XM_005270093.2
XP_005270150.1
1
662
XM_005270094.2
XP_005270151.1
1
662
XM_005270095.1
XP_005270152.1
1
662
XM_005270096.2
XP_005270153.1
1
662
XM_005270100.1
XP_005270157.1
1
662
XM_005270101.2
XP_005270158.1
1
662
XM_005270102.1
XP_005270159.1
1
662
XM_005270103.1
XP_005270160.1
1
662
XM_005270104.1
XP_005270161.1
1
662
XM_011540109.1
XP_011538411.1
1
662
XM_011540110.1
XP_011538412.1
1
662
XM_011540111.1
XP_011538413.1
1
662
XM_011540113.2
XP_011538415.1
1
662
XM_011540116.1
XP_011538418.1
1
662
XM_017016584.1
XP_016872073.1
1
662
XM_017016585.1
XP_016872074.1
1
662
XM_017016586.1
XP_016872075.1
1
662
XM_017016587.1
XP_016872076.1
1
662
XM_017016588.1
XP_016872077.1
1
662
XM_017016589.1
XP_016872078.1
1
662
XM_017016590.1
XP_016872079.1
1
662
XM_017016591.1
XP_016872080.1
1
662
XM_017016592.1
XP_016872081.1
1
662
XM_017016593.1
XP_016872082.1
1
662
XM_017016594.1
XP_016872083.1
1
662
XM_017016595.1
XP_016872084.1
1
662
XM_017016596.1
XP_016872085.1
1
662
GenBank mRNA
AB440195.1
-
1
124
AB451266.1
-
1
124
AK225809.1
-
1
635
AK299295.1
-
1
161
AK315243.1
-
1
297
BC032656.1
-
1
220
5533185
CR536574.1
-
1
124
FJ010166.1
-
1
217
FJ010167.1
-
1
217
FJ010168.1
-
1
221
FJ010169.1
-
1
221
FJ010170.1
-
1
217
FJ010171.1
-
1
161
FJ010172.1
-
1
221
FJ010174.1
-
1
579
HM352838.1
-
1
124
HM352839.1
-
1
124
HM352840.1
-
1
124
HM352841.1
-
1
124
HM352842.1
-
1
124
HM352843.1
-
1
124
HM352844.1
-
1
124
HM352845.1
-
1
124
HM352846.1
-
1
124
HM352847.1
-
1
124
HM352848.1
-
1
124
HM352849.1
-
1
124
HM352850.1
-
1
124
HM352851.1
-
1
124
Y11306.2
-
1
431
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
blood vessel development
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
cell cycle arrest
Wnt signaling pathway, calcium modulating pathway
cell proliferation
response to glucose
positive regulation of heparan sulfate proteoglycan biosynthetic process
pancreas development
positive regulation of insulin secretion
positive regulation of protein binding
regulation of hormone metabolic process
glucose homeostasis
negative regulation of sequence-specific DNA binding transcription factor activity
maintenance of DNA repeat elements
canonical Wnt signaling pathway involved in positive regulation of epithelial to mesenchymal transition
fat cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of protein export from nucleus
myoblast fate commitment
regulation of smooth muscle cell proliferation
positive regulation of protein kinase B signaling
canonical Wnt signaling pathway
negative regulation of canonical Wnt signaling pathway
beta-catenin-TCF complex assembly
negative regulation of type B pancreatic cell apoptotic process
negative regulation of extrinsic apoptotic signaling pathway
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II repressing transcription factor binding
transcription factor activity, sequence-specific DNA binding
protein binding
beta-catenin binding
transcription factor binding
protein kinase binding
nuclear hormone receptor binding
sequence-specific DNA binding
transcription regulatory region DNA binding
gamma-catenin binding
armadillo repeat domain binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-8jdkq:80/100.66.75.107:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0