Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008489.2 | NP_032515.2 | 3 | 536 | ||
GenBank mRNA | AK143562.1 | - | 3 | 537 | ||
AK158105.1 | - | 3 | 419 | |||
BC004795.1 | - | 3 | 424 | 3589072 | ||
X99347.1 | - | 3 | 486 |