Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other LPL SIRNA Assays ›
          Gene Symbol:
          Lpl
          Gene Name:
          lipoprotein lipase
          Gene Aliases:
          -
          Chromosome Location:
          Chr.8: 68880555 - 68906932 on Build GRCm38
          UniGene ID:
          Mm.1514
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 62800

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Lpl
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008509.2 NP_032535.2 2 359
          GenBank mRNA AK002645.1 - 2 285
          AK017272.1 - 357
          AK045064.1 - 2 248
          AK079446.1 - 2 248
          AK086023.1 - 2 180
          AK089473.1 - 2 258
          AK150328.1 - 2 360
          AK150355.1 - 2 360
          AK150375.1 - 2 358
          AK150457.1 - 2 255
          AK150488.1 - 2 360
          AK150505.1 - 2 360
          AK150532.1 - 2 360
          AK150577.1 - 2 360
          AK150585.1 - 2 360
          AK150593.1 - 2 360
          AK150646.1 - 2 357
          AK150672.1 - 2 278
          AK150691.1 - 2 360
          AK150984.1 - 2 360
          AK150990.1 - 2 360
          AK151006.1 - 2 360
          AK151038.1 - 2 258
          AK151049.1 - 2 360
          AK151093.1 - 2 360
          AK151099.1 - 2 360
          AK151105.1 - 2 360
          AK151112.1 - 2 361
          AK151195.1 - 2 258
          AK151216.1 - 2 361
          AK151369.1 - 2 278
          AK151434.1 - 2 278
          AK151521.1 - 2 278
          AK151540.1 - 2 248
          AK151563.1 - 2 360
          AK151630.1 - 2 258
          AK151679.1 - 2 360
          AK151698.1 - 2 360
          AK151727.1 - 2 360
          AK151772.1 - 2 360
          AK151774.1 - 2 258
          AK151801.1 - 2 248
          AK151828.1 - 2 248
          AK151866.1 - 2 360
          AK151870.1 - 2 360
          AK151872.1 - 2 360
          AK151893.1 - 2 248
          AK152014.1 - 2 360
          AK152035.1 - 2 360
          AK152049.1 - 2 360
          AK152053.1 - 2 248
          AK152079.1 - 2 360
          AK152238.1 - 2 360
          AK152251.1 - 2 357
          AK152350.1 - 2 360
          AK152390.1 - 2 199
          AK152400.1 - 2 258
          AK152481.1 - 2 361
          AK152657.1 - 2 360
          AK152706.1 - 2 361
          AK152733.1 - 2 258
          AK152775.1 - 2 360
          AK152869.1 - 2 360
          AK152956.1 - 2 360
          AK153006.1 - 2 360
          AK153148.1 - 2 360
          AK153242.1 - 2 361
          AK153425.1 - 2 360
          AK153428.1 - 2 360
          AK159268.1 - 2 360
          AK170486.1 - 2 360
          BC003305.1 - 2 348 3584788
          BC158040.1 - 2 193 9088156
          CT010344.1 - 2 161
          J03302.1 - 2 137
          M65258.1 - 2 321

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          lipase

          Gene Ontology Categories:

          Function(s) Process(es)

          lipid metabolic process
          fatty acid biosynthetic process
          positive regulation of macrophage derived foam cell differentiation
          positive regulation of cholesterol storage
          positive regulation of sequestering of triglyceride
          lipid catabolic process
          triglyceride biosynthetic process
          triglyceride catabolic process
          very-low-density lipoprotein particle remodeling
          triglyceride homeostasis
          lipoprotein lipase activity
          triglyceride lipase activity
          receptor binding
          protein binding
          heparin binding
          hydrolase activity
          triglyceride binding
          apolipoprotein binding
          carboxylic ester hydrolase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0