Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Anapc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008569.2 | NP_032595.2 | 19 | 2619 | ||
GenBank mRNA | AK052404.1 | - | 19 | 2600 | ||
AK054305.1 | - | 19 | 2601 | |||
AK077847.1 | - | 19 | 2620 | |||
AK085014.1 | - | 2 | 2612 | |||
AK090134.1 | - | 19 | 2601 | |||
AK161161.1 | - | 6 | 727 | |||
BC139002.1 | - | 19 | 2600 | 8862024 | ||
BC145492.1 | - | 19 | 2600 | 9054055 | ||
X80169.1 | - | 19 | 2604 |