Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other MSH2 SIRNA Assays ›
          Gene Symbol:
          Msh2
          Gene Name:
          mutS homolog 2 (E. coli)
          Gene Aliases:
          AI788990
          Chromosome Location:
          Chr.17: 87672557 - 87723713 on Build GRCm38
          UniGene ID:
          Mm.4619
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 63168

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Msh2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008628.2 NP_032654.1 4 795
          GenBank mRNA AK051960.1 - 4 792
          AK157842.1 - 4 808
          BC047117.1 - 4 803 4951173
          BC050897.1 - 4 778 6401835
          U21011.1 - 4 812
          X81143.1 - 4 795

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          DNA metabolism protein

          Gene Ontology Categories:

          Function(s) Process(es)

          meiotic mismatch repair
          in utero embryonic development
          somatic recombination of immunoglobulin genes involved in immune response
          oxidative phosphorylation
          DNA repair
          mismatch repair
          postreplication repair
          double-strand break repair
          meiotic gene conversion
          cellular response to DNA damage stimulus
          cell cycle
          cell cycle arrest
          reciprocal meiotic recombination
          germ cell development
          determination of adult lifespan
          male gonad development
          intrinsic apoptotic signaling pathway in response to DNA damage
          response to X-ray
          response to UV-B
          somatic hypermutation of immunoglobulin genes
          somatic recombination of immunoglobulin gene segments
          B cell mediated immunity
          B cell differentiation
          intra-S DNA damage checkpoint
          intrinsic apoptotic signaling pathway in response to DNA damage by p53 class mediator
          negative regulation of neuron apoptotic process
          maintenance of DNA repeat elements
          negative regulation of reciprocal meiotic recombination
          isotype switching
          negative regulation of DNA recombination
          positive regulation of helicase activity
          nucleotide binding
          magnesium ion binding
          four-way junction DNA binding
          Y-form DNA binding
          heteroduplex DNA loop binding
          double-strand/single-strand DNA junction binding
          DNA binding
          damaged DNA binding
          double-stranded DNA binding
          single-stranded DNA binding
          ATP binding
          protein C-terminus binding
          DNA-dependent ATPase activity
          ATPase activity
          centromeric DNA binding
          enzyme binding
          protein kinase binding
          mismatched DNA binding
          guanine/thymine mispair binding
          single base insertion or deletion binding
          dinucleotide insertion or deletion binding
          single guanine insertion binding
          single thymine insertion binding
          dinucleotide repeat insertion binding
          oxidized purine DNA binding
          MutLalpha complex binding
          protein homodimerization activity
          ADP binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0