Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pak3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195046.1 | NP_001181975.1 | 7 | 683 | ||
NM_001195047.1 | NP_001181976.1 | 6 | 638 | |||
NM_001195048.1 | NP_001181977.1 | 7 | 793 | |||
NM_001195049.1 | NP_001181978.1 | 9 | 1057 | |||
NM_008778.3 | NP_032804.2 | 6 | 666 | |||
GenBank mRNA | AF082297.1 | - | 5 | 580 | ||
AJ496262.1 | - | 5 | 548 | |||
AJ496263.1 | - | 6 | 593 | |||
AK031853.1 | - | 7 | 674 | |||
AK158893.1 | - | 7 | 793 | |||
AM943853.1 | - | 6 | 611 | |||
AM943854.1 | - | 7 | 656 | |||
BC053403.1 | - | 6 | 673 | 30060082 | ||
U39738.1 | - | 10 | 1064 |