Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nptx1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008730.2 | NP_032756.1 | 5 | 1888 | ||
GenBank mRNA | BC093479.1 | - | 5 | 1813 | 30947054 | |
BC139018.1 | - | 5 | 1795 | 8862040 | ||
BC139020.1 | - | 5 | 1795 | 8862042 |