Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Who We Serve
    • Biotech
    • Biopharma
    • CDMO
    • Lab Diagnostics
    • Industrial and Applied Sciences
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Aspire Member Program
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other PKD2 SIRNA Assays ›
          Gene Symbol:
          Pkd2
          Gene Name:
          polycystic kidney disease 2
          Gene Aliases:
          C030034P18Rik, PC2, TRPP2
          Chromosome Location:
          Chr.5: 104457224 - 104505819 on Build GRCm38
          UniGene ID:
          Mm.483692
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 63644

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Pkd2
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008861.3 NP_032887.3 1 495
          GenBank mRNA AF014010.1 - 1 398
          AK053502.1 - 1 400
          AK128961.1 - 1 377
          AK164673.1 - 1 386
          BC053058.1 - 1 502
          BC062969.1 - 1 502
          BU705556.1 - 1 501
          FJ609779.1 - 1 411
          Y13278.1 - 1 411
          Y14120.1 - 1 161

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001191934.

          Panther Classification:

          Molecular Function -

          ion channel

          Gene Ontology Categories:

          Function(s) Process(es)

          kidney development
          liver development
          embryonic placenta development
          heart looping
          detection of nodal flow
          transport
          ion transport
          calcium ion transport
          cellular calcium ion homeostasis
          cell cycle arrest
          positive regulation of cytosolic calcium ion concentration
          JAK-STAT cascade
          determination of left/right symmetry
          heart development
          positive regulation of gene expression
          regulation of cAMP metabolic process
          positive regulation of inositol 1,4,5-trisphosphate-sensitive calcium-release channel activity
          positive regulation of cyclin-dependent protein serine/threonine kinase activity involved in G1/S transition of mitotic cell cycle
          sodium ion transmembrane transport
          regulation of cell proliferation
          cytoplasmic sequestering of transcription factor
          positive regulation of nitric oxide biosynthetic process
          positive regulation of transcription from RNA polymerase II promoter
          detection of mechanical stimulus
          release of sequestered calcium ion into cytosol
          transmembrane transport
          negative regulation of ryanodine-sensitive calcium-release channel activity
          placenta blood vessel development
          renal tubule morphogenesis
          calcium ion transmembrane transport
          positive regulation of cell cycle arrest
          cellular response to hydrostatic pressure
          cellular response to osmotic stress
          cellular response to fluid shear stress
          potassium ion transmembrane transport
          determination of liver left/right asymmetry
          renal system development
          regulation of calcium ion import
          negative regulation of G1/S transition of mitotic cell cycle
          receptor binding
          voltage-gated ion channel activity
          voltage-gated calcium channel activity
          voltage-gated sodium channel activity
          calcium channel activity
          potassium channel activity
          calcium ion binding
          protein binding
          cytoskeletal protein binding
          channel activity
          voltage-gated cation channel activity
          identical protein binding
          protein homodimerization activity
          actinin binding
          HLH domain binding
          ion channel binding
          metal ion binding
          calcium-induced calcium release activity
          ATPase binding
          phosphoprotein binding
          muscle alpha-actinin binding
          alpha-actinin binding

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Consumer Health Data Privacy Policy Consumer Health Data Privacy Policy
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Notice
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          United States flag icon
          United States

          Your items have has been added!


          Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
          git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.43.0-2026.01.05-1.0