Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g2c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008868.3 | NP_032894.2 | 5 | 585 | ||
GenBank mRNA | BC029347.1 | - | 4 | 430 | 4234596 | |
CA464794.1 | - | 5 | 588 | |||
U18119.1 | - | 4 | 411 |