siRNA Details
Gene Symbol: Pou3f3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008900.2 | NP_032926.2 | 1 | 1301 | ||
GenBank mRNA | BC079869.1 | - | 1554 |

Gene Symbol: Pou3f3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008900.2 | NP_032926.2 | 1 | 1301 | ||
GenBank mRNA | BC079869.1 | - | 1554 |