Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmb6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008946.4 | NP_032972.3 | 4, 5 | 477 | ||
GenBank mRNA | AK078437.1 | - | 4, 5 | 449 | ||
AK132042.1 | - | 4, 5 | 460 | |||
AK167123.1 | - | 4, 5 | 449 | |||
AK167227.1 | - | 4, 5 | 445 | |||
BC013897.1 | - | 4, 5 | 437 | 4038758 | ||
U13393.1 | - | 4, 5 | 398 |