Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Cell Analysis
    • Antibodies
    • Mass Spectrometry
    • Cell Culture
    • Laboratory Instruments
    • Clinical and Diagnostics
    • Chromatography
    • Laboratory Equipment
    • Laboratory Supplies
    • Molecular Biology and Nucleic Acid Analysis
    • Sequence-Specific Nucleic Acid Products
    • See all product categories
  • Applications
    • Cell Culture and Transfection
    • Flow Cytometry
    • Cancer Research
    • Chromatography
    • Sequencing
    • PCR
    • Lab Solutions
    • Allergy Diagnostics
    • See all applications and techniques
  • Services
    • Cell Biology Services
    • Custom Services
    • Training Services
    • Enterprise Services
    • Lab Informatics Services
    • Financial and Leasing Services
    • Partnering and Licensing Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Food Safety Inspection Services
    • See all services
  • Help and Support
    • How to Order
    • Promotions and Online Offers
    • Contact Us
    • Change Location
    • Create a New Account
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Special Offers
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Custom Products & Projects
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other INPP5K SIRNA Assays ›
          Gene Symbol:
          Inpp5k
          Gene Name:
          inositol polyphosphate 5-phosphatase K
          Gene Aliases:
          C62, Pps, SKIP
          Chromosome Location:
          Chr.11: 75630507 - 75648865 on Build GRCm38
          UniGene ID:
          Mm.1458
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 63964

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Inpp5k
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_008916.2 NP_032942.1 10, 11 1330
          GenBank mRNA AF483522.1 - 10, 11 1250
          AF483523.1 - 10, 11 1250
          AK052817.1 - 9, 10 2074
          AK054436.1 - 10, 11 1349
          AK078104.1 - 10, 11 1341
          AK164290.1 - 10, 11 1319
          BC014725.1 - 10, 11 1260 3969089
          BC034549.1 - 2 2603
          BC066112.1 - 10, 11 1330 30361046
          U96724.1 - 10, 11 1361

          Back To Top

          More Information


          Other Species Targeted

          This siRNA also targets the following transcripts in Rat: NM_001013859, BC083572, FQ228522.

          Panther Classification:

          Molecular Function -

          phosphatase

          Gene Ontology Categories:

          Function(s) Process(es)

          in utero embryonic development
          negative regulation of protein phosphorylation
          regulation of glycogen biosynthetic process
          negative regulation of protein kinase activity
          G-protein coupled receptor signaling pathway
          negative regulation of peptidyl-threonine phosphorylation
          negative regulation of glucose transport
          dephosphorylation
          response to insulin
          cellular response to insulin stimulus
          cellular response to hormone stimulus
          negative regulation of peptidyl-serine phosphorylation
          negative regulation of dephosphorylation
          positive regulation of urine volume
          glucose homeostasis
          negative regulation of MAP kinase activity
          negative regulation by host of viral transcription
          negative regulation of glycogen biosynthetic process
          negative regulation of single stranded viral RNA replication via double stranded DNA intermediate
          negative regulation of transcription, DNA-templated
          positive regulation of transcription, DNA-templated
          negative regulation of insulin receptor signaling pathway
          inositol phosphate dephosphorylation
          phosphatidylinositol dephosphorylation
          negative regulation of stress fiber assembly
          negative regulation of protein kinase B signaling
          negative regulation of calcium ion transport
          cellular response to cAMP
          cellular response to tumor necrosis factor
          cellular response to epidermal growth factor stimulus
          protein targeting to plasma membrane
          negative regulation of protein targeting to membrane
          ruffle assembly
          negative regulation of glycogen (starch) synthase activity
          positive regulation of renal water transport
          inositol-polyphosphate 5-phosphatase activity
          vasopressin receptor activity
          inositol bisphosphate phosphatase activity
          hydrolase activity
          phosphatidylinositol-3,4,5-trisphosphate 5-phosphatase activity
          phosphatidylinositol trisphosphate phosphatase activity
          phosphatidylinositol phosphate 5-phosphatase activity
          inositol trisphosphate phosphatase activity
          inositol-1,4,5-trisphosphate 5-phosphatase activity
          inositol-1,3,4,5-tetrakisphosphate 5-phosphatase activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          • Social Media
          • Contact Us
          • Report a Site Issue
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          • Price & Freight Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Argentina flag icon
          Argentina

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-lffdw:80/100.66.79.31:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0