Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Cell Analysis
Antibodies
Mass Spectrometry
Cell Culture
Laboratory Instruments
Clinical and Diagnostics
Chromatography
Laboratory Equipment
Laboratory Supplies
Molecular Biology and Nucleic Acid Analysis
Sequence-Specific Nucleic Acid Products
See all product categories
Applications
Cell Culture and Transfection
Flow Cytometry
Cancer Research
Chromatography
Sequencing
PCR
Lab Solutions
Allergy Diagnostics
See all applications and techniques
Services
Cell Biology Services
Custom Services
Training Services
Enterprise Services
Lab Informatics Services
Financial and Leasing Services
Partnering and Licensing Services
360° CDMO and CRO Solutions
CDMO Services
CRO Services
Food Safety Inspection Services
See all services
Help and Support
How to Order
Promotions and Online Offers
Contact Us
Change Location
Create a New Account
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Special Offers
Contact Us
Quick Order
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Search All
Search
Search button
Contact Us
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Custom Products & Projects
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PTCH1 SIRNA Assays ›
Gene Symbol:
Ptch1
Gene Name:
patched homolog 1
Gene Aliases:
A230106A15Rik, Ptc, Ptc1, Ptch, mes
Chromosome Location:
Chr.13: 63508328 - 63573616 on Build GRCm38
UniGene ID:
Mm.228798
Species:
Mouse
Species Specific ID (Flybase ID):
-
siRNA ID
63999
Purification
Standard
HPLC
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
4,126.00
Your Price
Online offer:
4126.0
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: Ptch1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_008957.2
NP_032983.1
7
951
GenBank mRNA
AK147626.1
-
7
1160
U46155.1
-
7
951
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_053566, AF079162, AY357891.
Gene Ontology Categories:
Function(s)
Process(es)
negative regulation of transcription from RNA polymerase II promoter
branching involved in ureteric bud morphogenesis
in utero embryonic development
cell fate determination
neural tube formation
neural tube closure
heart morphogenesis
signal transduction
smoothened signaling pathway
regulation of mitotic cell cycle
pattern specification process
brain development
negative regulation of cell proliferation
epidermis development
regulation of smoothened signaling pathway
organ morphogenesis
dorsal/ventral pattern formation
epidermal cell fate specification
response to chlorate
positive regulation of cholesterol efflux
protein processing
spinal cord motor neuron differentiation
neural tube patterning
dorsal/ventral neural tube patterning
neural plate axis specification
embryonic limb morphogenesis
mammary gland development
regulation of protein localization
limb morphogenesis
hindlimb morphogenesis
regulation of growth
negative regulation of multicellular organism growth
regulation of cell proliferation
glucose homeostasis
negative regulation of sequence-specific DNA binding transcription factor activity
keratinocyte proliferation
positive regulation of epidermal cell differentiation
negative regulation of osteoblast differentiation
negative regulation of smoothened signaling pathway
embryonic organ development
negative regulation of epithelial cell proliferation
negative regulation of cell division
pharyngeal system development
mammary gland duct morphogenesis
mammary gland epithelial cell differentiation
smoothened signaling pathway involved in dorsal/ventral neural tube patterning
cell differentiation involved in kidney development
somite development
cellular response to cholesterol
cell proliferation involved in metanephros development
protein targeting to plasma membrane
patched binding
smoothened binding
protein binding
hedgehog receptor activity
heparin binding
zinc ion binding
cholesterol binding
cyclin binding
protein complex binding
hedgehog family protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.42.0-2026.01.03-1.0