siRNA Details
Gene Symbol: HOXA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000522.4 | NP_000513.2 | 2 | 1084 | ||
GenBank mRNA | BC075791.1 | - | 2 | 1084 | 30341785 |

Gene Symbol: HOXA13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000522.4 | NP_000513.2 | 2 | 1084 | ||
GenBank mRNA | BC075791.1 | - | 2 | 1084 | 30341785 |