Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rfx2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009056.2 | NP_033082.1 | 7 | 898 | ||
NM_027787.1 | NP_082063.1 | 8 | 973 | |||
GenBank mRNA | AK044439.1 | - | 8 | 973 | ||
BC004654.1 | - | 7 | 915 | 3497910 | ||
X76089.1 | - | 7 | 853 |