Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign In
Don't have an account ? Create Account
  • Applications
    • Real-Time PCR
    • Oligos, Primers, Probes and Genes
    • Cloning
    • Protein Biology
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Cell Analysis
    • Mass Spectrometry
    • Chromatography
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Online Order
    • Custom Primers & TaqMan Probes
    • miRNA Mimics & Inhibitors
    • Stealth RNAi / siRNA
    • Silencer Select siRNAs
    • Custom DNA Oligos
    • GeneArt Services
    • GeneArt Strings DNA Fragments
    • TrueGuide CRISPR gRNA
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • See all online orderable products
  • Services
    • Custom Services
    • Instrument Qualification Services
    • Technical Services
    • Enterprise Services
    • Pipette Services
    • Sample Request
    • Instrument Maintenance Services
    • Repairs and Relocation Services
    • OEM and Licensing Services
    • Events, Seminars, Training
    • See all services
  • Support
    • Catalogs
    • Application Notes
    • Safety Data Sheets
    • Legal and Regulatory
    • Certificates of Analysis Search
    • Nunc/Nalgene Product Certificates
    • e-learning
    • FAQs
    • Learning Centers
    • Technical Support Centers
    • See all help and support topics
  • Contact Us
  • Quick Order
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Contact Us
          • Quick Order
          • Sign In
            Sign In
            Don't have an account ? Create Account
            • Connect Your Lab
            • Custom Products & Projects
            • Instrument Management
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other CSF1R SIRNA Assays ›
          Gene Symbol:
          CSF1R
          Gene Name:
          colony stimulating factor 1 receptor
          Gene Aliases:
          C-FMS, CD115, CSF-1R, CSFR, FIM2, FMS, HDLS, M-CSF-R
          Chromosome Location:
          Chr.5: 150053291 - 150113372 on Build GRCh38
          UniGene ID:
          Hs.586219
          Species:
          Human
          Species Specific ID (Flybase ID):
          -
          siRNA ID 641

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price 4,126.00
          Your Price
          Online offer:
          4126.0
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: CSF1R
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_001288705.1 NP_001275634.1 3 740
          NM_005211.3 NP_005202.2 4 819
          NR_109969.1 3 740
          GenBank mRNA AB208819.1 - 3 690
          AK294467.1 - 3 655
          AK304491.1 - 3 521
          BC047521.1 - 3 722 4819527
          BC129939.1 - 3 660
          BX648599.1 - 2 610
          DR001419.1 - 3 616
          EU826593.1 - 3 527
          X03663.1 - 4 827

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          transmembrane signal receptor

          Gene Ontology Categories:

          Function(s) Process(es)

          positive regulation of protein phosphorylation
          inflammatory response
          signal transduction
          transmembrane receptor protein tyrosine kinase signaling pathway
          multicellular organism development
          axon guidance
          skeletal muscle tissue development
          cell proliferation
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          regulation of cell shape
          peptidyl-tyrosine phosphorylation
          cytokine-mediated signaling pathway
          olfactory bulb development
          forebrain neuron differentiation
          hemopoiesis
          monocyte differentiation
          macrophage differentiation
          osteoclast differentiation
          positive regulation of cell migration
          ruffle organization
          cellular response to macrophage colony-stimulating factor stimulus
          macrophage colony-stimulating factor signaling pathway
          positive regulation of tyrosine phosphorylation of Stat3 protein
          negative regulation of apoptotic process
          innate immune response
          regulation of bone resorption
          cell-cell junction maintenance
          positive regulation of osteoclast differentiation
          phosphatidylinositol metabolic process
          protein autophosphorylation
          phosphatidylinositol-mediated signaling
          mammary gland duct morphogenesis
          positive regulation of protein tyrosine kinase activity
          positive regulation of ERK1 and ERK2 cascade
          cellular response to cytokine stimulus
          positive regulation of protein serine/threonine kinase activity
          positive regulation of chemokine secretion
          positive regulation of cell motility
          regulation of actin cytoskeleton reorganization
          macrophage colony-stimulating factor receptor activity
          ATP binding
          protein phosphatase binding
          cytokine binding
          protein homodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Online Orderable Products
          • Privacy Policy for Online Ordering
          • Act on Specified Commercial Transactions
          • About Returns
          • About Displayed Prices
          • About Shipping Fees
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Study information disclosure
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Seminars
          • Blog
          About Thermo Fisher Plus Icon Minus Icon
          • Corporate Information
          • Social Responsibility (CSR)
          • Enhancing Customer Experience
          • Careers Careers
          • News
          • Trademarks
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Policy
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Japan flag icon
          Japan

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-ctdjb:80/100.66.76.55:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0