Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Resp18 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009049.1 | NP_033075.1 | 2, 3 | 292 | ||
GenBank mRNA | AK012477.1 | - | 2, 3 | 245 | ||
AK019440.1 | - | 2, 3 | 242 | |||
BC027535.1 | - | 2, 3 | 255 | 3376899 | ||
FJ621497.1 | - | 2, 3 | 228 | |||
L34214.1 | - | 2, 3 | 292 |