Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps17 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009092.3 | NP_033118.1 | 3 | 277 | ||
GenBank mRNA | AK007992.1 | - | 3 | 285 | ||
AK010647.1 | - | 3 | 279 | |||
AK012781.1 | - | 3 | 266 | |||
AK158902.1 | - | 2 | 343 | |||
AK162022.1 | - | 3 | 271 | |||
AK162335.1 | - | 3 | 269 | |||
BC081466.1 | - | 2 | 264 | 6412006 | ||
BC086900.1 | - | 3 | 270 | 6760363 | ||
BC086901.1 | - | 3 | 270 | 6763239 | ||
D25213.1 | - | 2 | 258 |