Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: St3gal3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161774.2 | NP_001155246.1 | 9, 10 | 983 | ||
NM_001285520.1 | NP_001272449.1 | 9, 10 | 1313 | |||
NM_001285521.1 | NP_001272450.1 | 8, 9 | 935 | |||
NM_009176.5 | NP_033202.3 | 9, 10 | 1327 | |||
GenBank mRNA | AK005053.1 | - | 9, 10 | 1272 | ||
AK013016.1 | - | 8, 9 | 874 | |||
AK136442.1 | - | 9, 10 | 1264 | |||
AK187627.1 | - | 1, 2 | 80 | |||
BC006710.1 | - | 9, 10 | 951 | 3584323 | ||
X84234.1 | - | 9, 10 | 806 |