Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Smarcc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_009211.2 | NP_033237.2 | 9 | 947 | ||
GenBank mRNA | AK042194.1 | - | 10 | 860 | ||
AK133922.1 | - | 9 | 922 | |||
AK143485.1 | - | 9 | 935 | |||
AK144122.1 | - | 9 | 921 | |||
AK144631.1 | - | 9 | 915 | |||
AK145352.1 | - | 9 | 914 | |||
BC052423.1 | - | 9 | 909 | 6400646 | ||
BC053064.1 | - | 9 | 903 | |||
BC056375.1 | - | 9 | 910 | |||
U85614.1 | - | 9 | 908 |