Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ? Create Account
  • Products
    • Antibodies
    • Cell Culture Media
    • Chemicals
    • Chromatography Columns and Cartridges
    • Lab Equipment
    • Lab Plasticware and Supplies
    • Microplates
    • Oligos, Primers, Probes and Genes
    • TaqMan Real-Time PCR Assays
    • Greener Products
    • See all product categories
  • Applications
    • Bioprocessing
    • Cell Culture and Transfection
    • Cell and Gene Therapy
    • Chromatography
    • Molecular Testing
    • Digital Solutions
    • DNA and RNA Extraction and Analysis
    • Spectroscopy, Elemental and Isotope Analysis
    • See all applications and techniques
  • Services
    • 360° CDMO and CRO Solutions
    • CDMO Services
    • CRO Services
    • Custom Services
    • Enterprise Services
    • Financial and Leasing Services
    • Instrument Services
    • Lab Informatics
    • OEM and Commercial Supply
    • Training Services
    • Unity Lab Services
    • See all services
  • Help and Support
    • How to Order
    • Instrument Support
    • Learning Centers
    • Register for an Account
    • Technical Support Centers
    • See all help and support topics
  • Popular
    • TaqMan Real-Time PCR Assays
      TaqMan Real-Time PCR Assays
    • Antibodies
      Antibodies
    • Oligos, Primers & Probes
      Oligos, Primers & Probes
    • GeneArt Gene Synthesis
      GeneArt Gene Synthesis
    • Cell Culture Plastics
      Cell Culture Plastics
  • Promotions
  • Contact Us
  • Quick Order
  • Order Status and Tracking
  • Documents and Certificates
Thermo Fisher Scientific Logo

Search

Search All
Search button
          • Order Status
          • Quick Order
          • Promos
          • Sign in
            Sign in
            Don't have an account ? Create Account
            • Account
            • Check Order Status
            • Connect: Lab, Data, Apps
            • Custom Products & Projects
            • Services Central
          This product has been added to your favorites list. Go to My Favorites

          System Message

          OKCancel
          LOADING ...
          See other SOX4 SIRNA Assays ›
          Gene Symbol:
          Sox4
          Gene Name:
          SRY (sex determining region Y)-box 4
          Gene Aliases:
          AA682046, Sox-4
          Chromosome Location:
          Chr.13: 28950716 - 28953682 on Build GRCm38
          UniGene ID:
          Mm.240627
          Species:
          Mouse
          Species Specific ID (Flybase ID):
          -
          siRNA ID 64697

          Purification
          Size
          Availability Inventoried
          Catalog # AM16708
          Price
          Your Price
          Online offer:
          Check your price ›
          • Genomic Map
          • siRNA Details
          • More Information

          Genomic Map

          LOADING...
          ×
          Back To Top

          siRNA Details

          Gene Symbol: Sox4
          Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
          RefSeq NM_009238.2 NP_033264.2 1 2031
          GenBank mRNA AK028989.1 - 2047
          AK031394.1 - 20
          AK051540.1 - 174
          AK053840.1 - 1 2049
          AK153771.1 - 30
          BC052736.1 - 1 2031 6822618
          BC060669.1 - 1 2152
          X70298.1 - 1 2026

          Back To Top

          More Information


          Panther Classification:

          Molecular Function -

          HMG box transcription factor

          Gene Ontology Categories:

          Function(s) Process(es)

          skeletal system development
          response to hypoxia
          neural tube formation
          pro-B cell differentiation
          mitral valve morphogenesis
          cardiac ventricle formation
          cardiac right ventricle morphogenesis
          atrial septum primum morphogenesis
          noradrenergic neuron differentiation
          transcription, DNA-templated
          regulation of transcription, DNA-templated
          transcription from RNA polymerase II promoter
          DNA damage response, signal transduction by p53 class mediator resulting in cell cycle arrest
          heart development
          positive regulation of cell proliferation
          negative regulation of cell proliferation
          glial cell proliferation
          spinal cord development
          spinal cord motor neuron differentiation
          glial cell development
          positive regulation of Wnt signaling pathway
          T cell differentiation
          endocrine pancreas development
          negative regulation of protein ubiquitination
          regulation of protein stability
          positive regulation of insulin secretion
          somatic stem cell population maintenance
          ascending aorta morphogenesis
          glucose homeostasis
          DNA damage response, detection of DNA damage
          positive regulation of apoptotic process
          positive regulation of translation
          positive regulation of transcription, DNA-templated
          positive regulation of transcription from RNA polymerase II promoter
          negative regulation of protein export from nucleus
          sympathetic nervous system development
          protein stabilization
          canonical Wnt signaling pathway
          limb bud formation
          ventricular septum morphogenesis
          negative regulation of cell death
          neuroepithelial cell differentiation
          kidney morphogenesis
          cellular response to glucose stimulus
          positive regulation of canonical Wnt signaling pathway
          positive regulation of N-terminal peptidyl-lysine acetylation
          transcription regulatory region sequence-specific DNA binding
          RNA polymerase II transcription factor activity, sequence-specific DNA binding
          core promoter sequence-specific DNA binding
          nucleic acid binding transcription factor activity
          transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
          RNA polymerase II transcription coactivator activity
          DNA binding
          transcription factor activity, sequence-specific DNA binding
          protein binding
          protein heterodimerization activity

          Back To Top

          Related Products

          • Anti-GAPDH, Mouse Monoclonal 6C5
          • Anti-β-Actin, Mouse Monoclonal AC-15
          • TaqMan® Gene Expression Cells-to-CT™
          • Neon™ Transfection System 100 µL Kit
          • Lipofectamine™ RNAiMAX Transfection Reagent
          • Invivofectamine 2.0
          • Lipofectamine™ 2000 Transfection Reagent
          • Neon™ Transfection System
          Ordering Plus Icon Minus Icon
          • Order Status
          • Order Help
          • Quick Order
          • Supply Center
          • eProcurement
          • Price & Freight Policy
          Support Plus Icon Minus Icon
          • Help and Support
          • Contact Us
          • Technical Support Centers
          • Documents and Certificates
          • Report a Site Issue
          Resources Plus Icon Minus Icon
          • Learning Centers
          • Promotions
          • Events and Webinars
          • Social Media
          About Thermo Fisher Plus Icon Minus Icon
          • About Us About Us
          • Careers Careers
          • Investors Investors
          • News News
          • Social Responsibility Social Responsibility
          • Trademarks
          • German Supply Chain Due Diligence Act
          • Policies and Notices
          Our Portfolio Plus Icon Minus Icon
          • Thermo Scientific
          • Applied Biosystems
          • Invitrogen
          • Gibco
          • Ion Torrent
          • Fisher Scientific
          • Unity Lab Services
          • Patheon
          • PPD
          • Terms & Conditions
          • Privacy Information Center
          • Legal Notice
          © 2006-2026 Thermo Fisher Scientific Inc. All rights reserved. All trademarks are the property of Thermo Fisher Scientific and its subsidiaries unless otherwise specified.
          Germany flag icon
          Germany

          Your items have has been added!


          Host server : magellan-search-blue-55ff9ddd88-vsgnr:80/100.66.75.107:80.
          git-commit: d366ff9721d93504b2ac26a183b2b3e3d0e7d9ec
          git-url: https://github.com/thermofisher/magellan-search
          git-branch: release/2.42.0-2026.01.03-1.0